Merck
RSG1 is required for cilia-dependent neural tube closure.
Genesis (New York, N.Y. : 2000) (2024-05-09)
David Engelhardt, Amber Marean, David McKean, Juliette Petersen, Lee Niswander
ABSTRACT
Cilia play a key role in the regulation of signaling pathways required for embryonic development, including the proper formation of the neural tube, the precursor to the brain and spinal cord. Forward genetic screens were used to generate mouse lines that display neural tube defects (NTD) and secondary phenotypes useful in interrogating function. We describe here the L3P mutant line that displays phenotypes of disrupted Sonic hedgehog signaling and affects the initiation of cilia formation. A point mutation was mapped in the L3P line to the gene Rsg1, which encodes a GTPase-like protein. The mutation lies within the GTP-binding pocket and disrupts the highly conserved G1 domain. The mutant protein and other centrosomal and IFT proteins still localize appropriately to the basal body of cilia, suggesting that RSG1 GTPase activity is not required for basal body maturation but is needed for a downstream step in axonemal elongation.
Sigma-Aldrich
Anti-PAX6 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-ISL-1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-SHH Antibody, clone 5H4, clone 5H4, from mouse