- POR overexpression induces tamoxifen-resistance in breast cancer through the STAT1/c-Myc pathway.
POR overexpression induces tamoxifen-resistance in breast cancer through the STAT1/c-Myc pathway.
Breast cancer is the most common cancer in women worldwide. Although tamoxifen (TAM), a selective estrogen receptor (ER) modulator, is widely used to treat ER-positive breast cancers, resistance to TAM remains a major clinical problem. NADPH-dependent cytochrome P450 reductase (POR) is known to participate in drug metabolism and steroid metabolism. Recent studies showed that high POR expression was correlated with poor outcomes in triple-negative breast cancer (TNBC), and POR might be a prognostic biomarker in TNBC. However, the role of POR in TAM resistance is still elusive. In this study, we found that high POR expression was associated with poor prognosis of ER-positive and TAM-treated breast cancer patients. In addition, COX analysis showed that POR expression was an independent prognostic biomarker for ER-positive as well as TAM-treated breast cancer patients. Furthermore, our results suggested that POR overexpression promoted TAM resistance by activating the STAT1/c-Myc pathway in ER-positive breast cancer cells. Immunohistochemical analysis showed that high POR/STAT1 expression was correlated with poor prognosis in TAM-treated breast cancer patients. Notably, combined treatment with TAM and a specific STAT1 inhibitor Fludarabine was more effective for inhibiting TAM-resistant breast cancer cells. Altogether, our findings suggested that POR overexpression induced TAM resistance through STAT1/c-Myc pathway and might serve as an independent prognostic biomarker in TAM-treated breast cancer patients. Combining TAM and STAT1 inhibitors might be an effective strategy for treating POR-induced TAM-resistant breast cancer.