Merck
  • MAPK- and AKT-activated thyroid cancers are sensitive to group I PAK inhibition.
MAPK- and AKT-activated thyroid cancers are sensitive to group I PAK inhibition.
Endocrine-related cancer (2019-05-31)
Christina M Knippler, Motoyasu Saji, Neel Rajan, Kyle Porter, Krista M D La Perle, Matthew D Ringel
ABSTRACT
The number of individuals who succumb to thyroid cancer has been increasing and those who are refractory to standard care have limited therapeutic options, highlighting the importance of developing new treatments for patients with aggressive forms of the disease. Mutational activation of MAPK signaling, through BRAF and RAS mutations and/or gene rearrangements, and activation of PI3K signaling, through mutational activation of PIK3CA or loss of PTEN, are well described in aggressive thyroid cancer. We previously reported overactivation and overexpression of p21-activated kinases (PAKs) in aggressive human thyroid cancer invasive fronts and determined that PAK1 functionally regulated thyroid cancer cell migration. We reported mechanistic crosstalk between the MAPK and PAK pathways that are BRAF-dependent but MEK independent, suggesting that PAK and MEK inhibition might be synergistic. In the present study, we tested this hypothesis. Pharmacologic inhibition of group I PAKs using two PAK kinase inhibitors, G-5555 or FRAX1036, reduced thyroid cancer cell viability, cell cycle progression and migration and invasion, with greater potency for G-5555. Combination of G-5555 with vemurafenib was synergistic in BRAFV600E-mutated thyroid cancer cell lines. Finally, G-5555 restrained thyroid size of BRAFV600E-driven murine papillary thyroid cancer by >50% (P < 0.0001) and reduced carcinoma formation (P = 0.0167), despite maintenance of MAPK activity. Taken together, these findings suggest both that group I PAKs may be a new therapeutic target for thyroid cancer and that PAK activation is functionally important for BRAFV600E-mediated thyroid cancer development.
Sigma-Aldrich
G-5555, ≥98% (HPLC)