Merck
Usnic acid, as a biotic factor, changes the ploidy level in mosses.
Ecology and evolution (2018-03-14)
Michal Goga, Dajana Ručová, Vladislav Kolarčik, Marko Sabovljević, Martin Bačkor, Ingeborg Lang
ABSTRACT
Lichens and mosses often share the same environmental conditions where they compete for substrate and other essential factors. Lichens use secondary metabolites as allelochemicals to repel surrounding plants and potential rivals. In mosses, endoreduplication leads to the occurrence of various ploidy levels in the same individual and has been suggested as an adaptation to abiotic stresses. Here, we show that also biotic factors such as usnic acid, an allelochemical produced by lichens, directly influenced the level of ploidy in mosses. Application of usnic acid changed the nuclei proportion and significantly enhanced the endoreduplication index in two moss species, Physcomitrella patens and Pohlia drummondii. These investigations add a new aspect on secondary metabolites of lichens which count as biotic factors and affect ploidy levels in mosses.
Sigma-Aldrich
(+)-Usnic acid, 98%
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPrice
OR54637-250MG250 mg
Ships in 14 days - from Aldrich Partner.
€126.90
OR54637-1G1 g
Ships in 14 days - from Aldrich Partner.
€299.60
To order products, please contact your local dealer.