Merck
  • Severe deficiency of the voltage-gated sodium channel NaV1.2 elevates neuronal excitability in adult mice.
Severe deficiency of the voltage-gated sodium channel NaV1.2 elevates neuronal excitability in adult mice.
Cell reports (2021-08-05)
Jingliang Zhang, Xiaoling Chen, Muriel Eaton, Jiaxiang Wu, Zhixiong Ma, Shirong Lai, Anthony Park, Talha S Ahmad, Zhefu Que, Ji Hea Lee, Tiange Xiao, Yuansong Li, Yujia Wang, Maria I Olivero-Acosta, James A Schaber, Krishna Jayant, Chongli Yuan, Zhuo Huang, Nadia A Lanman, William C Skarnes, Yang Yang
ABSTRACT
Scn2a encodes the voltage-gated sodium channel NaV1.2, a main mediator of neuronal action potential firing. The current paradigm suggests that NaV1.2 gain-of-function variants enhance neuronal excitability, resulting in epilepsy, whereas NaV1.2 deficiency impairs neuronal excitability, contributing to autism. However, this paradigm does not explain why ∼20%-30% of individuals with NaV1.2 deficiency still develop seizures. Here, we report the counterintuitive finding that severe NaV1.2 deficiency results in increased neuronal excitability. Using a NaV1.2-deficient mouse model, we show enhanced intrinsic excitability of principal neurons in the prefrontal cortex and striatum, brain regions known to be involved in Scn2a-related seizures. This increased excitability is autonomous and reversible by genetic restoration of Scn2a expression in adult mice. RNA sequencing reveals downregulation of multiple potassium channels, including KV1.1. Correspondingly, KV channel openers alleviate the hyperexcitability of NaV1.2-deficient neurons. This unexpected neuronal hyperexcitability may serve as a cellular basis underlying NaV1.2 deficiency-related seizures.
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
4-(Trifluoromethyl)-L-phenylglycine, ≥98.0%
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%