Merck
  • Inactivation of glycogen synthase kinase-3β (GSK-3β) enhances skeletal muscle oxidative metabolism.
Inactivation of glycogen synthase kinase-3β (GSK-3β) enhances skeletal muscle oxidative metabolism.
Biochimica et biophysica acta. Molecular basis of disease (2017-09-26)
W F Theeuwes, H R Gosker, R C J Langen, K J P Verhees, N A M Pansters, A M W J Schols, A H V Remels
ABSTRACT
Aberrant skeletal muscle mitochondrial oxidative metabolism is a debilitating feature of chronic diseases such as chronic obstructive pulmonary disease, type 2 diabetes and chronic heart failure. Evidence in non-muscle cells suggests that glycogen synthase kinase-3β (GSK-3β) represses mitochondrial biogenesis and inhibits PPAR-γ co-activator 1 (PGC-1), a master regulator of cellular oxidative metabolism. The role of GSK-3β in the regulation of skeletal muscle oxidative metabolism is unknown. We hypothesized that inactivation of GSK-3β stimulates muscle oxidative metabolism by activating PGC-1 signaling and explored if GSK-3β inactivation could protect against physical inactivity-induced alterations in skeletal muscle oxidative metabolism. GSK-3β was modulated genetically and pharmacologically in C2C12 myotubes in vitro and in skeletal muscle in vivo. Wild-type and muscle-specific GSK-3β knock-out (KO) mice were subjected to hind limb suspension for 14days. Key constituents of oxidative metabolism and PGC-1 signaling were investigated. In vitro, knock-down of GSK-3β increased mitochondrial DNA copy number, protein and mRNA abundance of oxidative phosphorylation (OXPHOS) complexes and activity of oxidative metabolic enzymes but also enhanced protein and mRNA abundance of key PGC-1 signaling constituents. Similarly, pharmacological inhibition of GSK-3β increased transcript and protein abundance of key constituents and regulators of mitochondrial energy metabolism. Furthermore, GSK-3β KO animals were protected against unloading-induced decrements in expression levels of these constituents. Inactivation of GSK-3β up-regulates skeletal muscle mitochondrial metabolism and increases expression levels of PGC-1 signaling constituents. In vivo, GSK-3β KO protects against inactivity-induced reductions in muscle metabolic gene expression.