Merck
  • Proteomic analysis reveals rotator cuff injury caused by oxidative stress.
Proteomic analysis reveals rotator cuff injury caused by oxidative stress.
Therapeutic advances in chronic disease (2021-04-03)
Tao Yuan, Hong Qian, Xin Yu, Jia Meng, Cheng-Teng Lai, Hui Jiang, Jian-Ning Zhao, Ni-Rong Bao
ABSTRACT
Rotator cuff tendinopathy is common and is related to pain and dysfunction. However, the pathological mechanism of rotator cuff injury and shoulder pain is unclear. Objective: to investigate the pathological mechanism of rotator cuff injury and shoulder pain, and screen out the marker proteins related to rotator cuff injury by proteomics. Subacromial synovium specimens were collected from patients undergoing shoulder arthroscopic surgery. The experimental group were patients with rotator cuff repair surgery, and the control group were patients with habitual dislocation of the shoulder joint. Pathological examination was performed, and then followed by non-labeled quantitative proteomic detection. Finally, from analysis of the biological information of the samples, specific proteins related to rotator cuff injury and shoulder pain were deduced by functional analysis of differential proteins. All the patients in experimental groups were representative. A large number of adipocytes and inflammatory cells were found in the pathological sections of the experimental group; the proteomics analysis screen identified 80 proteins with significant differences, and the analysis of protein function revealed that S100A11 (p = 0.011), PLIN4 (p = 0.017), HYOU1 (p = 0.002) and CLIC1 (p = 0.007) were closely related to oxidative stress and chronic inflammation. Rotator cuff injury is closely related to oxidative stress and chronic inflammatory response, and the results suggest that the expression of S100A11, PLIN4, HYOU1 and CLIC1 in the synovium of rotator cuff injury provides a new marker for the study of its pathological mechanism.
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Indole-3-acetic acid sodium salt, BioReagent, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Supelco
Empore SPE Disks, matrix active group C18, diam. 47 mm, pk of 20