Merck
  • Nitric oxide resets kisspeptin-excited GnRH neurons via PIP2 replenishment.
Nitric oxide resets kisspeptin-excited GnRH neurons via PIP2 replenishment.
Proceedings of the National Academy of Sciences of the United States of America (2021-01-15)
Stephanie Constantin, Daniel Reynolds, Andrew Oh, Katherine Pizano, Susan Wray
ABSTRACT
Fertility relies upon pulsatile release of gonadotropin-releasing hormone (GnRH) that drives pulsatile luteinizing hormone secretion. Kisspeptin (KP) neurons in the arcuate nucleus are at the center of the GnRH pulse generation and the steroid feedback control of GnRH secretion. However, KP evokes a long-lasting response in GnRH neurons that is hard to reconcile with periodic GnRH activity required to drive GnRH pulses. Using calcium imaging, we show that 1) the tetrodotoxin-insensitive calcium response evoked by KP relies upon the ongoing activity of canonical transient receptor potential channels maintaining voltage-gated calcium channels in an activated state, 2) the duration of the calcium response is determined by the rate of resynthesis of phosphatidylinositol 4,5-bisphosphate (PIP2), and 3) nitric oxide terminates the calcium response by facilitating the resynthesis of PIP2 via the canonical pathway guanylyl cyclase/3',5'-cyclic guanosine monophosphate/protein kinase G. In addition, our data indicate that exposure to nitric oxide after KP facilitates the calcium response to a subsequent KP application. This effect was replicated using electrophysiology on GnRH neurons in acute brain slices. The interplay between KP and nitric oxide signaling provides a mechanism for modulation of the refractory period of GnRH neurons after KP exposure and places nitric oxide as an important component for tonic GnRH neuronal pulses.
Sigma-Aldrich
Propylamine, purum, ≥99.0% (GC)
Supelco
Flufenamic acid, analytical standard, for drug analysis
Sigma-Aldrich
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one, powder
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Lanthanum(III) chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Luteinizing hormone releasing hormone, ≥97% (HPLC)
Sigma-Aldrich
8-(4-Chlorophenylthio)-guanosine 3′,5′-cyclic monophosphate sodium salt, ≥95% (HPLC), powder