Merck
  • Enhancement of myogenic potential of muscle progenitor cells and muscle healing during pregnancy.
Enhancement of myogenic potential of muscle progenitor cells and muscle healing during pregnancy.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2021-02-11)
Aiping Lu, Ping Guo, Haiying Pan, Chieh Tseng, Krishna M Sinha, Fan Yang, Alex Scibetta, Yan Cui, Matthieu Huard, Ling Zhong, Sudheer Ravuri, Johnny Huard
ABSTRACT
The decline of muscle regenerative potential with age has been attributed to a diminished responsiveness of muscle progenitor cells (MPCs). Heterochronic parabiosis has been used as a model to study the effects of aging on stem cells and their niches. These studies have demonstrated that, by exposing old mice to a young systemic environment, aged progenitor cells can be rejuvenated. One interesting idea is that pregnancy represents a unique biological model of a naturally shared circulatory system between developing and mature organisms. To test this hypothesis, we evaluated the muscle regeneration potential of pregnant mice using a cardiotoxin (CTX) injury mouse model. Our results indicate that the pregnant mice demonstrate accelerated muscle healing compared to nonpregnant control mice following muscle injury based on improved muscle histology, superior muscle regeneration, and a reduction in inflammation and necrosis. Additionally, we found that MPCs isolated from pregnant mice display a significant improvement of myogenic differentiation capacity in vitro and muscle regeneration in vivo when compared to the MPCs from nonpregnant mice. Furthermore, MPCs from nonpregnant mice display enhanced myogenic capacity when cultured in the presence of serum obtained from pregnant mice. Our proteomics data from these studies provides potential therapeutic targets to enhance the myogenic potential of progenitor cells and muscle repair.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
(Z)-4-Hydroxytamoxifen, ≥98% Z isomer
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
DAPI, for nucleic acid staining
Cy®3-Streptavidin, Cytiva PA43001