Merck
  • Sex-dimorphic effects of biogenesis of lysosome-related organelles complex-1 deficiency on mouse perinatal brain development.
Sex-dimorphic effects of biogenesis of lysosome-related organelles complex-1 deficiency on mouse perinatal brain development.
Journal of neuroscience research (2020-05-22)
Frank Y Lee, Jennifer Larimore, Victor Faundez, Esteban C Dell'Angelica, Cristina A Ghiani
ABSTRACT
The function(s) of the Biogenesis of Lysosome-related Organelles Complex-1 (BLOC-1) during brain development is to date largely unknown. Here, we investigated how its absence alters the trajectory of postnatal brain development using as model the pallid mouse. Most of the defects observed early postnatally in the mutant mice were more prominent in males than in females and in the hippocampus. Male mutant mice, but not females, had smaller brains as compared to sex-matching wild types at postnatal day 1 (P1), this deficit was largely recovered by P14 and P45. An abnormal cytoarchitecture of the pyramidal cell layer of the hippocampus was observed in P1 pallid male, but not female, or juvenile mice (P45), along with severely decreased expression levels of the radial glial marker Glutamate-Aspartate Transporter. Transcriptomic analyses showed that the overall response to the lack of functional BLOC-1 was more pronounced in hippocampi at P1 than at P45 or in the cerebral cortex. These observations suggest that absence of BLOC-1 renders males more susceptible to perinatal brain maldevelopment and although most abnormalities appear to have been resolved in juvenile animals, still permanent defects may be present, resulting in faulty neuronal circuits, and contribute to previously reported cognitive and behavioral phenotypes in adult BLOC-1-deficient mice.
Sigma-Aldrich
Anti-Reelin Antibody, a.a. 164-189 mreelin, clone 142, clone 142, Chemicon®, from mouse
Sigma-Aldrich
Anti-Internexin α Antibody, serum, Chemicon®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-GLAST Antibody, clone 8C11.1, clone 8C11.1, from mouse