Merck
  • Quantitative Investigation of the Process Parameters of Electrohydrodynamic Direct-Writing and Their Effects on Fiber Surface Roughness and Cell Adhesion.
Quantitative Investigation of the Process Parameters of Electrohydrodynamic Direct-Writing and Their Effects on Fiber Surface Roughness and Cell Adhesion.
Polymers (2020-10-30)
Chen Jiang, Kan Wang, Xuzhou Jiang, Chuck Zhang, Ben Wang
ABSTRACT
Electrohydrodynamic (EHD) direct-writing has been widely used to fabricate micro/nanofibers that can serve as a building block in tissue engineering scaffolds. However, the application of EHD direct-writing in tissue engineering is limited by the lack of fundamental knowledge in the correlations among the process parameters, the fiber surface roughness, and the cell adhesion performance. Without a standardized experimental setting and the quantitative database, inconsistent results have been reported. Here, we quantitatively investigate the process-structure-property relationships as the first step towards a better understanding of the EHD direct-writing technology for tissue engineering. Polycaprolactone (PCL) solution is used as a model ink material, and human mesenchymal stem cells (hMSCs) are used to study cell adhesion on PCL fibers. We investigate the different jetting modes defined by the applied voltage, the feed rate, and the nozzle-collector distance. The quantitative effects of process parameters on the fiber surface roughness and the cell adhesion performance are experimentally determined. The quantitative process-structure-property relationships revealed in this study provide guidelines for controlling the surface roughness and the cell adhesion performance of EHD direct-written fibers. This study will facilitate the application of EHD direct-writing in tissue engineering.
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Polycaprolactone, average Mn 80,000