Merck
  • Cell Plasticity-Related Phenotypes and Taxanes Resistance in Castration-Resistant Prostate Cancer.
Cell Plasticity-Related Phenotypes and Taxanes Resistance in Castration-Resistant Prostate Cancer.
Frontiers in oncology (2020-11-24)
Natalia Jiménez, Òscar Reig, Ruth Montalbo, Maria Milà-Guasch, Lluis Nadal-Dieste, Giancarlo Castellano, Juan José Lozano, Iván Victoria, Albert Font, Alejo Rodriguez-Vida, Joan Carles, Cristina Suárez, Montserrat Domènech, Núria Sala-González, Pedro Luis Fernández, Leonardo Rodríguez-Carunchio, Sherley Díaz, Aleix Prat, Mercedes Marín-Aguilera, Begoña Mellado
ABSTRACT
The prostatic tumor cells plasticity is involved in resistance to hormone-therapy, allowing these cells to survive despite androgen receptor inhibition. However, its role in taxanes resistance has not been fully established. Gene expression of plasticity-related phenotypes such as epithelial-mesenchymal transition (EMT), stem cell-like and neuroendocrine (NE) phenotypes was studied in vitro, in silico, in circulating tumor cells (CTCs) (N=22) and in tumor samples (N=117) from taxanes-treated metastatic castration-resistant prostate cancer (mCRPC) patients. Docetaxel (D)-resistant cells presented a more pronounced EMT phenotype than cabazitaxel (CZ)-resistant cells. In silico analysis revealed ESRP1 down-regulation in taxane-exposed mCRPC samples. Cell plasticity-related changes occurred in CTCs after taxanes treatment. Tumor EMT phenotype was associated with lower PSA progression-free survival (PFS) to D (P<0.001), and better to CZ (P=0.002). High ESRP1 expression was independently associated with longer PSA-PFS (P<0.001) and radiologic-PFS (P=0.001) in D and shorter PSA-PFS in the CZ cohort (P=0.041). High SYP expression was independently associated with lower PSA-PFS in D (P=0.003) and overall survival (OS) in CZ (P=0.002), and high EZH2 expression was associated with adverse OS in D-treated patients (P=0.013). In conclusion, EMT profile in primary tumor is differentially associated with D or CZ benefit and NE dedifferentiation correlates with adverse taxanes clinical outcome.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
25 g
Please contact Customer Service for Availability
€44.80
100 g
Please contact Customer Service for Availability
€132.00
To order products, please contact your local dealer.
Sigma-Aldrich
Anti-ESRP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
25 g
Please contact Customer Service for Availability
€44.80
100 g
Please contact Customer Service for Availability
€132.00
To order products, please contact your local dealer.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
25 g
Please contact Customer Service for Availability
€44.80
100 g
Please contact Customer Service for Availability
€132.00
To order products, please contact your local dealer.
Sigma-Aldrich
Monoclonal Anti-CHGA antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0166, purified immunoglobulin, buffered aqueous glycerol solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
25 g
Please contact Customer Service for Availability
€44.80
100 g
Please contact Customer Service for Availability
€132.00
To order products, please contact your local dealer.