Merck
  • Low temperature bacterial expression of the neutral amino acid transporters SLC1A5 (ASCT2), and SLC6A19 (B0AT1).
Low temperature bacterial expression of the neutral amino acid transporters SLC1A5 (ASCT2), and SLC6A19 (B0AT1).
Molecular biology reports (2020-08-11)
Michele Galluccio, Marta Pantanella, Deborah Giudice, Stefania Brescia, Cesare Indiveri
ABSTRACT
It is well established that Escherichia coli represents a powerful tool for the over-expression of human proteins for structure/function studies. In many cases, such as for membrane transporters, the bacterial toxicity or the aggregation of the target protein hamper the expression limiting the application of this tool. The aim of this study was finding the appropriate conditions for the expression of reluctant proteins that is the human neutral amino acid transporters ASCT2 and B0AT1, that have great relevance to human health in cancer therapy and in COVID-19 research, respectively. The cDNAs coding for the proteins of interest were cloned in the pCOLD I vector and different E. coli strains (BL21 codon plus RIL, and RosettaGami2) were cultured in absence or in presence of glucose (0.5-1%), at low temperature (15 °C), and low inducer concentrations (10-100 µM). Cell growth and protein production were monitored by optical density measurements and western blotting assay, respectively. Even though in different conditions, the expression of both amino acid transporters was obtained.Reducing the growth rate of specific E. coli strains by lowering the temperature and the IPTG concentration, together with the addition of glucose, two reluctant human neutral amino acid transporters have been expressed in E. coli. The results have a potentially great interest in drug discovery since ASCT2 is an acknowledged target of anticancer therapy, and B0AT1 together with ACE2 is part of a receptor for the SARS-Cov-2 RBD proteins.
Sigma-Aldrich
Human Dermal Fibroblasts: HDF, adult
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
Sigma-Aldrich
Anti-polyHistidine−Peroxidase antibody, Mouse monoclonal, clone HIS-1, purified from hybridoma cell culture