Merck
  • The Hippo Effector Transcriptional Coactivator with PDZ-Binding Motif Cooperates with Oncogenic β-Catenin to Induce Hepatoblastoma Development in Mice and Humans.
The Hippo Effector Transcriptional Coactivator with PDZ-Binding Motif Cooperates with Oncogenic β-Catenin to Induce Hepatoblastoma Development in Mice and Humans.
The American journal of pathology (2020-04-14)
Shu Zhang, Jie Zhang, Katja Evert, Xiaolei Li, Pin Liu, Andras Kiss, Zsuzsa Schaff, Cindy Ament, Yi Zhang, Monica Serra, Matthias Evert, Nianyong Chen, Feng Xu, Xin Chen, Junyan Tao, Diego F Calvisi, Antonio Cigliano
ABSTRACT
Hepatoblastoma (HB) is the most common pediatric liver tumor. Though Wnt/β-catenin and Hippo cascades are implicated in HB development, studies on crosstalk between β-catenin and Hippo downstream effector transcriptional coactivator with PDZ-binding motif (TAZ) in HB are lacking. Expression levels of TAZ and β-catenin in human HB specimens were assessed by immunohistochemistry. Functional interplay between TAZ and β-catenin was determined by overexpression of an activated form of TAZ (TAZS89A), either alone or combined with an oncogenic form of β-catenin (ΔN90-β-catenin), in mouse liver via hydrodynamic transfection. Activation of TAZ often co-occurred with that of β-catenin in clinical specimens. Although the overexpression of TAZS89A alone did not induce hepatocarcinogenesis, concomitant overexpression of TAZS89A and ΔN90-β-catenin triggered the development of HB lesions exhibiting both epithelial and mesenchymal features. Mechanistically, TAZ/β-catenin-driven HB development required TAZ interaction with transcriptional enhanced associate domain factors. Blockade of the Notch cascade did not inhibit TAZ/β-catenin-dependent HB formation in mice but suppressed the mesenchymal phenotype. Neither Yes-associated protein nor heat shock factor 1 depletion affected HB development in TAZ/β-catenin mice. In human HB cell lines, silencing of TAZ resulted in decreased cell growth, which was further reduced when TAZ knockdown was associated with suppression of either β-catenin or Yes-associated protein. Overall, our study identified TAZ as a crucial oncogene in HB development and progression.
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Monoclonal Anti-WWTR1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0371, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%