Merck
  • Feeder-free derivation of induced pluripotent stem cells from adult human adipose stem cells.
Feeder-free derivation of induced pluripotent stem cells from adult human adipose stem cells.
Proceedings of the National Academy of Sciences of the United States of America (2009-10-07)
Ning Sun, Nicholas J Panetta, Deepak M Gupta, Kitchener D Wilson, Andrew Lee, Fangjun Jia, Shijun Hu, Athena M Cherry, Robert C Robbins, Michael T Longaker, Joseph C Wu
ABSTRACT
Ectopic expression of transcription factors can reprogram somatic cells to a pluripotent state. However, most of the studies used skin fibroblasts as the starting population for reprogramming, which usually take weeks for expansion from a single biopsy. We show here that induced pluripotent stem (iPS) cells can be generated from adult human adipose stem cells (hASCs) freshly isolated from patients. Furthermore, iPS cells can be readily derived from adult hASCs in a feeder-free condition, thereby eliminating potential variability caused by using feeder cells. hASCs can be safely and readily isolated from adult humans in large quantities without extended time for expansion, are easy to maintain in culture, and therefore represent an ideal autologous source of cells for generating individual-specific iPS cells.
Sigma-Aldrich
Anti-TRA-1-81 Antibody, clone TRA-1-81, clone TRA-1-81, Chemicon®, from mouse
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, Alexa Fluor 488 Conjugate, clone TRA - 1-60, from mouse, ALEXA FLUOR 488
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, clone TRA-1-60, Chemicon®, from mouse