Merck
  • Sulforaphane sensitizes human breast cancer cells to paclitaxel-induced apoptosis by downregulating the NF-κB signaling pathway.
Sulforaphane sensitizes human breast cancer cells to paclitaxel-induced apoptosis by downregulating the NF-κB signaling pathway.
Oncology letters (2017-06-11)
Seung-Hyun Kim, Hyun-Jung Park, Dong-Oh Moon
ABSTRACT
Sulforaphane (SFN), an isothiocyanate present in cruciferous vegetables, has been demonstrated to inhibit the growth of various types of cancer cell. The aim of the present study was to investigate whether SFN sensitizes breast cancer cells to paclitaxel-induced apoptosis and to identify the signal pathway through which SFN mediates apoptosis. Combined treatment of breast cancer cells with SFN and paclitaxel resulted in increased activation of apoptotic signaling pathway members, including caspase-3, -8 and -9, and cytochrome c, compared with treatment with SFN or paclitaxel alone. In addition, treatment with SFN and paclitaxel resulted in downregulation of the nuclear factor kappa B signaling pathway, and reduced protein expression of apoptosis regulator Bcl-2 and phosphorylated AKT serine/threonine kinase. Furthermore, SFN-paclitaxel-induced apoptosis was inhibited by overexpression of Bcl-2. The results of the present study suggest that combined treatment with SFN and paclitaxel is a novel therapeutic strategy for the treatment of breast cancer.
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Anti-Bcl-2 Antibody, clone 1D7.2, clone 1D7.2, from rat
Sigma-Aldrich
Anti-Caspase 9 Antibody, proform & lg. subunit, Chemicon®, from rabbit