Merck
  • Lysosomal integral membrane protein-2 (LIMP-2/SCARB2) is involved in lysosomal cholesterol export.
Lysosomal integral membrane protein-2 (LIMP-2/SCARB2) is involved in lysosomal cholesterol export.
Nature communications (2019-08-08)
Saskia Heybrock, Kristiina Kanerva, Ying Meng, Chris Ing, Anna Liang, Zi-Jian Xiong, Xialian Weng, Young Ah Kim, Richard Collins, William Trimble, Régis Pomès, Gilbert G Privé, Wim Annaert, Michael Schwake, Joerg Heeren, Renate Lüllmann-Rauch, Sergio Grinstein, Elina Ikonen, Paul Saftig, Dante Neculai
ABSTRACT
The intracellular transport of cholesterol is subject to tight regulation. The structure of the lysosomal integral membrane protein type 2 (LIMP-2, also known as SCARB2) reveals a large cavity that traverses the molecule and resembles the cavity in SR-B1 that mediates lipid transfer. The detection of cholesterol within the LIMP-2 structure and the formation of cholesterol-like inclusions in LIMP-2 knockout mice suggested the possibility that LIMP2 transports cholesterol in lysosomes. We present results of molecular modeling, crosslinking studies, microscale thermophoresis and cell-based assays that support a role of LIMP-2 in cholesterol transport. We show that the cavity in the luminal domain of LIMP-2 can bind and deliver exogenous cholesterol to the lysosomal membrane and later to lipid droplets. Depletion of LIMP-2 alters SREBP-2-mediated cholesterol regulation, as well as LDL-receptor levels. Our data indicate that LIMP-2 operates in parallel with Niemann Pick (NPC)-proteins, mediating a slower mode of lysosomal cholesterol export.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human NPC1
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
Sigma-Aldrich
MISSION® esiRNA, targeting human SCARB2
Sigma-Aldrich
Sandoz 58-035, >98% (HPLC), powder
Avanti
PhotoClick Cholesterol, Hex-5′-ynyl 3β-hydroxy-6-diazirinyl-5α-cholan-24-oate, powder