Merck
  • Cholesterol Metabolism Is a Druggable Axis that Independently Regulates Tau and Amyloid-β in iPSC-Derived Alzheimer's Disease Neurons.
Cholesterol Metabolism Is a Druggable Axis that Independently Regulates Tau and Amyloid-β in iPSC-Derived Alzheimer's Disease Neurons.
Cell stem cell (2019-01-29)
Rik van der Kant, Vanessa F Langness, Cheryl M Herrera, Daniel A Williams, Lauren K Fong, Yves Leestemaker, Evelyne Steenvoorden, Kevin D Rynearson, Jos F Brouwers, J Bernd Helms, Huib Ovaa, Martin Giera, Steven L Wagner, Anne G Bang, Lawrence S B Goldstein
ABSTRACT
Genetic, epidemiologic, and biochemical evidence suggests that predisposition to Alzheimer's disease (AD) may arise from altered cholesterol metabolism, although the molecular pathways that may link cholesterol to AD phenotypes are only partially understood. Here, we perform a phenotypic screen for pTau accumulation in AD-patient iPSC-derived neurons and identify cholesteryl esters (CE), the storage product of excess cholesterol, as upstream regulators of Tau early during AD development. Using isogenic induced pluripotent stem cell (iPSC) lines carrying mutations in the cholesterol-binding domain of APP or APP null alleles, we found that while CE also regulate Aβ secretion, the effects of CE on Tau and Aβ are mediated by independent pathways. Efficacy and toxicity screening in iPSC-derived astrocytes and neurons showed that allosteric activation of CYP46A1 lowers CE specifically in neurons and is well tolerated by astrocytes. These data reveal that CE independently regulate Tau and Aβ and identify a druggable CYP46A1-CE-Tau axis in AD.
Sigma-Aldrich
Fatostatin hydrobromide, ≥98% (HPLC), powder
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
FTI-277 trifluoroacetate salt, ≥95% (HPLC), film
Sigma-Aldrich
K-604, ≥98% (HPLC)
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Anti-Tau Antibody, clone Tau 7, clone Tau 7, from mouse
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal (751-770) Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Rosuvastatin calcium, ≥98% (HPLC)
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
Anti-τ (Tau) antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Anti-Histone H3 Antibody, 0.5 mg/mL, Upstate®
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®