Merck
  • Two short-acting kappa opioid receptor antagonists (zyklophin and LY2444296) exhibited different behavioral effects from the long-acting antagonist norbinaltorphimine in mouse anxiety tests.
Two short-acting kappa opioid receptor antagonists (zyklophin and LY2444296) exhibited different behavioral effects from the long-acting antagonist norbinaltorphimine in mouse anxiety tests.
Neuroscience letters (2016-01-19)
Peng Huang, Tatyana Yakovleva, Jane V Aldrich, Julia Tunis, Christopher Parry, Lee-Yuan Liu-Chen
ABSTRACT
Prototypical long-acting kappa opioid receptor (KOPR) antagonists [e.g., norbinaltorphimine (norBNI)] have been reported to exert anxiolytic-like effects in several commonly used anxiety tests in rodents including the novelty-induced hypophagia (NIH) and elevated plus maze (EPM) tests. It remains unknown if the short-acting KOPR antagonists (e.g., zyklophin and LY2444296) have similar effects. In this study effects of zyklophin and LY2444296 (s.c.) were investigated in the NIH and EPM tests in mice 1h post-injection and compared with norBNI (i.p.) 48h post-administration. In the NIH test, zyklophin at 3 and 1mg/kg, but not 0.3mg/kg, or LY2444296 at 30mg/kg decreased the latency of palatable food consumption in novel cages, but had no effect in training cages, similar to norBNI (10mg/kg). Zyklophin at 3 or 1mg/kg increased or had a trend of increasing the amount of palatable food consumption in novel cages, with no effects in training cages, further indicating its anxiolytic-like effect, but norBNI (10mg/kg) and LY2444296 (30mg/kg) did not. In the EPM test, norBNI (10mg/kg) increased open arm time and % open arm entries or time, but zyklophin at all three doses and LY2444296 (30mg/kg) had no effects. In addition, zyklophin at 3mg/kg increased numbers of close and total arm entries on EPM, suggesting increased activity; however, norBNI and LY2444296 had no effects on close and total arm entries. Thus, all three KOPR antagonists had anxiolytic-like effects in the NIH test. However, only the long-acting one (norBNI), but not the short-acting ones (zyklophin and LY2444296), demonstrated anti-anxiety like effects in the EPM test. It remains to be investigated if the differences are due to the differences in their durations of action and/or pharmacodynamic properties.
Sigma-Aldrich
LY2444296, ≥98% (HPLC)