Merck
  • Aster Proteins Facilitate Nonvesicular Plasma Membrane to ER Cholesterol Transport in Mammalian Cells.
Aster Proteins Facilitate Nonvesicular Plasma Membrane to ER Cholesterol Transport in Mammalian Cells.
Cell (2018-09-18)
Jaspreet Sandhu, Shiqian Li, Louise Fairall, Simon G Pfisterer, Jennifer E Gurnett, Xu Xiao, Thomas A Weston, Dipti Vashi, Alessandra Ferrari, Jose L Orozco, Celine L Hartman, David Strugatsky, Stephen D Lee, Cuiwen He, Cynthia Hong, Haibo Jiang, Laurent A Bentolila, Alberto T Gatta, Tim P Levine, Annie Ferng, Richard Lee, David A Ford, Stephen G Young, Elina Ikonen, John W R Schwabe, Peter Tontonoz
ABSTRACT
The mechanisms underlying sterol transport in mammalian cells are poorly understood. In particular, how cholesterol internalized from HDL is made available to the cell for storage or modification is unknown. Here, we describe three ER-resident proteins (Aster-A, -B, -C) that bind cholesterol and facilitate its removal from the plasma membrane. The crystal structure of the central domain of Aster-A broadly resembles the sterol-binding fold of mammalian StARD proteins, but sequence differences in the Aster pocket result in a distinct mode of ligand binding. The Aster N-terminal GRAM domain binds phosphatidylserine and mediates Aster recruitment to plasma membrane-ER contact sites in response to cholesterol accumulation in the plasma membrane. Mice lacking Aster-B are deficient in adrenal cholesterol ester storage and steroidogenesis because of an inability to transport cholesterol from SR-BI to the ER. These findings identify a nonvesicular pathway for plasma membrane to ER sterol trafficking in mammals.
Product Description
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
LG100268, ≥98% (HPLC)
Sigma-Aldrich
25-Hydroxycholesterol, ≥98%
Sigma-Aldrich
Methyl-β-cyclodextrin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
22(R)-Hydroxycholesterol, ≥98%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Avanti
NBD-6 Cholesterol, Avanti Research - A Croda Brand 810251P, powder