Merck
  • Hybridization dependent cleavage of internally modified disulfide-peptide nucleic acids.
Hybridization dependent cleavage of internally modified disulfide-peptide nucleic acids.
Bioorganic & medicinal chemistry letters (2005-01-25)
Iris Boll, Roland Krämer, Andriy Mokhir
ABSTRACT
Selectivity of the cleavage of single stranded over hybridized forms of internally modified disulfide-peptide nucleic acids (PNA) has been optimized using a series of phosphines and thiols, which have different sizes and charges. For the most selective cleaver found (tris-(carboxyethyl)-phosphine), reactivity of single stranded PNA is 33 times higher than that of the PNA-DNA duplex. Selectivity of single stranded disulfide-PNA cleavage has been explained in terms of electrostatic interaction between the substrate and the cleaver.
Sigma-Aldrich
4-(Dimethylamino)phenyldiphenylphosphine, 95%
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99%
Sigma-Aldrich
Cysteamine, ~95%
Sigma-Aldrich
3-Mercaptopropionic acid, ≥99.0% (HPLC)