Merck
  • Sulforaphane prevents angiotensin II-induced cardiomyopathy by activation of Nrf2 via stimulating the Akt/GSK-3ß/Fyn pathway.
Sulforaphane prevents angiotensin II-induced cardiomyopathy by activation of Nrf2 via stimulating the Akt/GSK-3ß/Fyn pathway.
Redox biology (2018-01-22)
Ying Xin, Yang Bai, Xin Jiang, Shanshan Zhou, Yuehui Wang, Kupper A Wintergerst, Taixing Cui, Honglei Ji, Yi Tan, Lu Cai
ABSTRACT
Activation of nuclear factor erythroid 2-related factor 2 (Nrf2) by sulforaphane (SFN) protects from, and deletion of the Nrf2 gene exaggerates, diabetic cardiomyopathy. Angiotensin II (Ang II) plays a critical role in the development of diabetic cardiomyopathy. Therefore, whether SFN prevents Ang II-induced cardiomyopathy through activation of Nrf2 was examined using wild-type, global deletion of Nrf2 gene (Nrf2-KO) and cardiomyocyte-specific overexpression of Nrf2 gene (Nrf2-TG) mice. Administration of a subpressor dose of Ang II to wild-type mice induced cardiac oxidative stress, inflammation, remodeling and dysfunction, all of which could be prevented by SFN treatment with Nrf2 up-regulation and activation. Nrf2-KO mice are susceptible, and Nrf2-TG mice are resistant, respectively, to Ang II-induced cardiomyopathy. Meanwhile, the ability of SFN to protect against Ang II-induced cardiac damage was lost in Nrf2-KO mice. Up-regulation and activation of Nrf2 by SFN is accompanied by activation of Akt, inhibition of glycogen synthase kinase (GSK)-3β, and accumulation of Fyn in nuclei. In vitro up-regulation of Nrf2 by SFN was abolished and nuclear Fyn accumulation was increased when cardiac cells were exposed to a PI3K inhibitor or GSK-3β-specific activator. These results suggest that Nrf2 plays a central role in the prevention of Ang II-induced cardiomyopathy, and SFN prevents Ang II-induced cardiomyopathy partially via the Akt/GSK-3β/Fyn-mediated Nrf2 activation.
Sigma-Aldrich
Nuclei Isolation Kit: Nuclei PURE Prep, sufficient for 15 nuclei preparations (~1-10×107 cells or 1g of tissue per preparation)
Sigma-Aldrich
3-Nitro-L-tyrosine, crystalline