
Valproic Acid Related Compound A

Synonym(s):
2-Allyl-4-pentenoic acid, Diallylacetic acid
Empirical Formula (Hill Notation):
C8H12O2
CAS Number:
Molecular Weight:
140.18
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2