![Valinomycin Ready Made Solution, ~1 mg/mL in DMSO](/deepweb/assets/sigmaaldrich/product/structures/340/996/c229afe2-6017-4366-8005-9a1c20231e08/640/c229afe2-6017-4366-8005-9a1c20231e08.png)
Valinomycin
![Valinomycin Ready Made Solution, ~1 mg/mL in DMSO](/deepweb/assets/sigmaaldrich/product/structures/340/996/c229afe2-6017-4366-8005-9a1c20231e08/640/c229afe2-6017-4366-8005-9a1c20231e08.png)
Synonym(s):
Cyclo(L-Val-D-HyIva-D-Val-L-Lac-)3: HyIva = α-Hydroxyisovaleric acid, Lac = Lactic acid
Empirical Formula (Hill Notation):
C54H90N6O18
CAS Number:
Molecular Weight:
1111.32
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-3 of 3
Compare
Product Number
Description
Pricing