
Valaciclovir impurity D

Synonym(s):
2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl N-ethyl-L-valinate, N-Ethyl-L-valine 2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester
Empirical Formula (Hill Notation):
C15H24N6O4
CAS Number:
Molecular Weight:
352.39
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing