
Tris-Borate-EDTA buffer

Synonym(s):
TBE buffer
Products
Product Category
Brand
Feature
pH Value
Physical Form
Product Line
Research Area
Quality Segment
Application
Sterilization Method
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing