
Trifluralin

Synonym(s):
2,6-Dinitro-N,N-dipropyl-4-trifluoromethylaniline
Empirical Formula (Hill Notation):
C13H16F3N3O4
CAS Number:
Molecular Weight:
335.28
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2