![Tetracycline hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/150/108/3539faca-a7b3-46da-b270-1db181b25504/640/3539faca-a7b3-46da-b270-1db181b25504.png)
Tetracycline hydrochloride
![Tetracycline hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/150/108/3539faca-a7b3-46da-b270-1db181b25504/640/3539faca-a7b3-46da-b270-1db181b25504.png)
Empirical Formula (Hill Notation):
C22H24N2O8 · HCl
CAS Number:
Molecular Weight:
480.90
Products
Product Category
Brand
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Activity Spectrum
Agency
Format
Quality Segment
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing