
Super-Hydride® solution

Synonym(s):
Lithium triethylborohydride solution
Linear Formula:
Li(C2H5)3BH
CAS Number:
Molecular Weight:
105.94
Products
Brand
Formula Weight
Physical Form
Quality Segment
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing