
Sodium acetate-13C2

Synonym(s):
Acetic acid-13C2 sodium salt
Linear Formula:
13CH313CO2Na
CAS Number:
Molecular Weight:
84.02
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing