
(S)-(+)-Arginine hydrochloride

Synonym(s):
L-Arginine monohydrochloride, (S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride
Empirical Formula (Hill Notation):
C6H14N4O2 · HCl
CAS Number:
Molecular Weight:
210.66
Products
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing