Screw cap tubes without caps, conical bottom, self-standing
Synonym(s):
Screw cap tubes without caps, conical bottom, self-standing
Products
Product Category
Color
Manufacturer
Sterility
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Screw cap tubes without caps, conical bottom, self-standing, size 2.0 mL, amber, pkg of 8x500caps/cs
Screw cap tubes without caps, conical bottom, self-standing, size 1.5 mL, amber, pkg of 8x500caps/cs
Screw cap tubes without caps, conical bottom, self-standing, size 2.0 mL, clear, pkg of 8x500caps/cs
Screw cap tubes without caps, conical bottom, self-standing, size 0.5 mL, amber, pkg of 8x500caps/cs