
Rufinamide

Synonym(s):
1-[(2,6-difluorophenyl)methyl]-1H-1,2,3-Triazole-4-carboxamide
Empirical Formula (Hill Notation):
C10H8F2N4O
CAS Number:
Molecular Weight:
238.19
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing