
Pyrroloquinoline quinone

Synonym(s):
4,5-Dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid, Methoxatin, PQQ
Empirical Formula (Hill Notation):
C14H6N2O8
CAS Number:
Molecular Weight:
330.21
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing