
Procyanidin B1

Synonym(s):
(−)-Epicatechin (4β-8)-(+)-catechin, cis,trans′′-4,8′′-Bi-(3,3′,4′,5,7-Pentahydroxyflavane), Proanthocyanidin B1
Empirical Formula (Hill Notation):
C30H26O12
CAS Number:
Molecular Weight:
578.52
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing