
Poly(ethylene glycol-ran-propylene glycol) monobutyl ether

Linear Formula:
CH3(CH2)3(OCH2CH2)x[OCH2CH(CH3)]yOH
CAS Number:
Products
Boiling Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing