
Pimobendan

Synonym(s):
4,5-Dihydro-6-[2-(4-methoxyphenyl)-1H-benzimidazol-6-yl]-5-methyl-3(2H)-pyridazinone, Pimobendan
Empirical Formula (Hill Notation):
C19H18N4O2
CAS Number:
Molecular Weight:
334.37
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4