Skip to Content
Merck
Oxymorphone solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Oxymorphone solution

Oxymorphone solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®
Synonym(s):
Oxymorphone solution
Empirical Formula (Hill Notation):
C17H19NO4
CAS Number:
Molecular Weight:
301.34

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique