Skip to Content
Merck
Octisalate Pharmaceutical Secondary Standard; Certified Reference Material

Octisalate

Octisalate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2-Ethylhexyl salicylate, Octisalate, Octyl salicylate
Linear Formula:
(HO)C6H4CO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
250.33
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique