
Norethisterone

Synonym(s):
19-Norethindrone, 17α-Ethynyl-19-nortestosterone, 17-Hydroxy-19-nor-17α-4-pregnen-20-yn-3-one, 17α-Ethynyl-17β-hydroxy-19-nor-4-androsten-3-one, 19-Nor-17α-ethynyl-4-androsten-17β-ol-3-one, 19-Norethisterone
Empirical Formula (Hill Notation):
C20H26O2
CAS Number:
Molecular Weight:
298.42
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing