n-Heptyl β-D-thioglucopyranoside
Synonym(s):
Heptyl thioglucoside, Heptyl-β-D-1-thioglucopyranoside
Empirical Formula (Hill Notation):
C13H26O5S
CAS Number:
Molecular Weight:
294.41
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing