
Metaraminol bitartrate

Synonym(s):
Metaraminol (+)-bitartrate salt, (−)-m-Hydroxyphenylpropanolamine bitartrate salt
Empirical Formula (Hill Notation):
C9H13NO2 · C4H6O6
CAS Number:
Molecular Weight:
317.29
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2