Skip to Content
BT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
meta-Phosphoric acid
Synonym(s):
Metaphosphoric acid
Linear Formula:
(HPO
3
)
n
EC Number:
253-433-4
Products
Product Category
inorganic acids (1)
Brand
Sigma-Aldrich (1)
Physical Form
Product Line
Chemical Composition
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.00546
meta-Phosphoric acid,
pieces for analysis (stabilized with sodium metaphosphate) EMSURE
®
Expand
View Pricing
04103
meta-Phosphoric acid,
100%, lumps (glassy)
Expand
View Pricing