
Manganese sulfate monohydrate

Synonym(s):
Manganese(II) sulfate monohydrate
Linear Formula:
MnSO4 · H2O
Products
Product Category
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing