Skip to Content
Merck
Magnesium carbonate hydroxide hydrate 99%

Magnesium carbonate hydroxide hydrate

Magnesium carbonate hydroxide hydrate 99%
Empirical Formula (Hill Notation):
C4Mg4O12 · H2MgO2 · xH2O
Molecular Weight:
395.58 (anhydrous basis)
EC Number:

Products

Product Category
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing