![MacConkey Broth Purple suitable for microbiology, NutriSelect® Plus](/deepweb/assets/sigmaaldrich/product/images/306/167/77b79fd0-3a9d-4115-bb3c-924a8e4c2a4e/640/77b79fd0-3a9d-4115-bb3c-924a8e4c2a4e.jpg)
MacConkey Broth Purple
![MacConkey Broth Purple suitable for microbiology, NutriSelect® Plus](/deepweb/assets/sigmaaldrich/product/images/306/167/77b79fd0-3a9d-4115-bb3c-924a8e4c2a4e/640/77b79fd0-3a9d-4115-bb3c-924a8e4c2a4e.jpg)
Synonym(s):
MacConkey Bromocresol purple Broth, MacConkey G Broth
Products
Product Category
Brand
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing