
Lisinopril for peak identification

Synonym(s):
Lisinopril, (S)-1-[N2-(1-carboxy-3-phenylpropyl)-lysyl-proline dihydrate, MK-521
Empirical Formula (Hill Notation):
C21H31O5N3 · 2H2O
CAS Number:
Molecular Weight:
441.52
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing