
L-Asparagine-13C4 monohydrate

Synonym(s):
(S)-2-Aminosuccinic acid 4-amide-13C4 monohydrate, (S)·-·(+)·-·2-·Aminosuccinamic acid-13C4, (S)-2-Aminosuccinic acid 4-amide-13C4 monohydrate, L-Aspartic acid 4-amide-13C4 monohydrate, 13C Labeled asparagine
Linear Formula:
H2N13CO13CH213CH(NH2)13CO2H · H2O
CAS Number:
Molecular Weight:
154.10
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing