
Iodine monochloride solution

Synonym(s):
Chloroiodide solution, Wijs solution
Linear Formula:
ICl
CAS Number:
Molecular Weight:
162.36
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing