
Imatinib impurity H

Synonym(s):
3-Acetylpyridine, Methyl 3-pyridyl ketone
Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing